Anti-IDO2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09548
EVBIEB09548EA 335 EUR
EVBIEB09548
Anti-IDO2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IDO2 (indoleamine 2,3-dioxygenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR