Anti-DNAJC6 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB09621
EVBIEB09621EA 202 EUR
EVBIEB09621
Anti-DNAJC6 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNAJC6 (DnaJ (Hsp40) homolog, subfamily C, member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR