Anti-POU4F2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB10032
EVBIEB10032EA 339 EUR
EVBIEB10032
Anti-POU4F2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: POU4F2 (POU class 4 homeobox 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR