Anti-ALDH3A2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB10131
EVBIEB10131EA 339 EUR
EVBIEB10131
Anti-ALDH3A2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH3A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Dog, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR