Anti-ETFB Goat Polyclonal Antibody (Biotinylated)
Lieferant: Everest Biotech
EB10172-B
EVBIEB10172-BEA 335 EUR
EVBIEB10172-B
Anti-ETFB Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: ETFB
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Cow