Anti-NPAS4 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB10571
EVBIEB10571EA 339 EUR
EVBIEB10571
Anti-NPAS4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NPAS4 (neuronal PAS domain protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR