Anti-CD28 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB10741
EVBIEB10741EA 339 EUR
EVBIEB10741
Anti-CD28 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: CD28 (CD28 molecule)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR