Anti-Delta-Sarcoglycan Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB10758
EVBIEB10758EA 335 EUR
EVBIEB10758
Anti-Delta-Sarcoglycan Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: Delta-Sarcoglycan
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR