Anti-PI15 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11365
EVBIEB11365EA 335 EUR
EVBIEB11365
Anti-PI15 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PI15 (peptidase inhibitor 15)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR