Anti-Loki / Chk2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11423
EVBIEB11423EA 202 EUR
EVBIEB11423
Anti-Loki / Chk2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Loki / Chk2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Drosophila
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR