Anti-MUC5B Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11765
EVBIEB11765EA 335 EUR
EVBIEB11765
Anti-MUC5B Goat Polyclonal Antibody
Antikörper
Western Blot: Not yet tested - our routinely used western blotting protocol does not allow detection of proteins as large as the calculated size of 515kDa according to NP_083077.2. Therefore we cannot recommend an optimal concentration and the antibody i

Type: Primary
Antigen: MUC5B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR