Anti-DNAJC5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11845
EVBIEB11845EA 335 EUR
EVBIEB11845
Anti-DNAJC5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DNAJC5 (DnaJ (Hsp40) homolog, subfamily C, member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR