Anti-ZNF697 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11883
EVBIEB11883EA 194 EUR
EVBIEB11883
Anti-ZNF697 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZNF697 (zinc finger protein 697)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Bovine, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR