Anti-AIM2 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11894
EVBIEB11894EA 194 EUR
EVBIEB11894
Anti-AIM2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: AIM2 (absent in melanoma 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR