Anti-MIA / CD-RAP Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB11934
EVBIEB11934EA 335 EUR
EVBIEB11934
Anti-MIA / CD-RAP Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MIA / CD-RAP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR