Anti-FABP3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12000
EVBIEB12000EA 335 EUR
EVBIEB12000
Anti-FABP3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FABP3 (fatty acid binding protein 3, muscle and heart (mammary-derived growth inhibitor))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR