Anti-PLK5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12016
EVBIEB12016EA 335 EUR
EVBIEB12016
Anti-PLK5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PLK5 (polo-like kinase 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR