Anti-BEND5 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB12141
EVBIEB12141EA 335 EUR
EVBIEB12141
Anti-BEND5 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: BEND5 (BEN domain containing 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR