Anti-S100B Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12316
EVBIEB12316EA 335 EUR
EVBIEB12316
Anti-S100B Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: S100B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR