Anti-CA12 Goat Polyclonal Antibody (Biotinylated)

Lieferant: Everest Biotech
EB12320-B
EVBIEB12320-BEA 339 EUR
EVBIEB12320-B
Anti-CA12 Goat Polyclonal Antibody (Biotinylated)
Antikörper
Type: Primary
Antigen: CA12
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR