Anti-FGA Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12376
EVBIEB12376EA 339 EUR
EVBIEB12376
Anti-FGA Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 75kDa band observed in Human Platelets lysates (calculated MW of 69.8kDa according to NP_068657.1). Recommended concentration: 0.01-0.03µg/ml.

Type: Primary
Antigen: FGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR