Anti-Adig Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12487
EVBIEB12487EA 202 EUR
EVBIEB12487
Anti-Adig Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Adig
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR