Anti-CD74 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12552
EVBIEB12552EA 339 EUR
EVBIEB12552
Anti-CD74 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 33kDa band observed in Human Tonsil lysates (calculated MW of 33.5kDa according to NP_001020330.1). Recommended concentration: 0.01-0.03µg/ml.

Type: Primary
Antigen: CD74
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR