Anti-CD3E Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12592
EVBIEB12592EA 339 EUR
EVBIEB12592
Anti-CD3E Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 22kDa band observed in lysates of cell lines Jurkat and Molt4 (calculated MW of 23.1kDa according to NP_000724.1). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR