Anti-CD3G Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12594
EVBIEB12594EA 339 EUR
EVBIEB12594
Anti-CD3G Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 23kDa band observed in lysates of cell line Jurkat (calculated MW of 20.5kDa according to NP_000064.1). Recommended concentration: 0.3-1µg/ml.

Type: Primary
Antigen: CD3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR