Anti-CD247 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12595
EVBIEB12595EA 335 EUR
EVBIEB12595
Anti-CD247 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 16+17kDa band observed in Human Tonsil lysates and in lysates of cell line Jurkat (calculated MW of 18.7kDa according to NP_932170.1). Recommended concentration: 0.03-0.1µg/ml.

Type: Primary
Antigen: CD247
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR