Anti-CAPRIN1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12638
EVBIEB12638EA 339 EUR
EVBIEB12638
Anti-CAPRIN1 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 80+140kDa bands observed in lysates of cell line Caco-2, HeLa, Jurkat and K562 (calculated MW of 78.4kDa according to NP_005889.3). The ~140kDa band corresponds to the reported p137 (Ellis and Luzio, J Biol Chem. 1995 Sep 1;270(35):2

Type: Primary
Antigen: CAPRIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Rat, Dog, Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR