Anti-CFD Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12706
EVBIEB12706EA 339 EUR
EVBIEB12706
Anti-CFD Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 30kDa band observed in Human Adipose lysates (calculated MW of 27kDa according to NP_001919.2). Recommended concentration: 1-3µg/ml.

Type: Primary
Antigen: CFD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Pig, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR