Anti-HDAC3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12818
EVBIEB12818EA 335 EUR
EVBIEB12818
Anti-HDAC3 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 50kDa band observed in nuclear lysates of cell line Jurkat (calculated MW of 48.8kDa according to NP_003874.2). Recommended concentration: 0.1-0.3µg/ml.

Type: Primary
Antigen: HDAC3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR