Anti-HSPE1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12835
EVBIEB12835EA 339 EUR
EVBIEB12835
Anti-HSPE1 Goat Polyclonal Antibody
Antikörper
Western Blot: Approx 10kDa band observed in lysates of cell line HepG2 (calculated MW of 10.9kDa according to NP_002148.1). Recommended concentration: 0.01-0.03µg/ml.

Type: Primary
Antigen: HSPE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR