Anti-NLRP5 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12894
EVBIEB12894EA 339 EUR
EVBIEB12894
Anti-NLRP5 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NLRP5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR