Anti-OAZ1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB12909
EVBIEB12909EA 200 EUR
EVBIEB12909
Anti-OAZ1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: OAZ1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Dog, Pig, Cow
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR