Anti-CD84 Rabbit Polyclonal Antibody (FITC)

Lieferant: G-Biosciences
ITT5256-100U-F
GENOITT5256-100U-FEA 936 EUR
GENOITT5256-100U-F
Anti-CD84 Rabbit Polyclonal Antibody (FITC)
Antikörper
Type: Primary
Antigen: CD84
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR