Anti-His tag Mouse Monoclonal Antibody

Lieferant: Genscript
A00186-100
GENSA00186-100EA 640 EUR
GENSA00186-100
Anti-His tag Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: His tag (His tag)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR