Anti-VSV-G-tag Rabbit Polyclonal Antibody

Lieferant: Genscript
A00199-40
GENSA00199-40EA 173 EUR
GENSA00199-40
Anti-VSV-G-tag Rabbit Polyclonal Antibody
Antikörper
Type: Tag
Antigen: VSV-G-tag
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR