Anti-His tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Genscript
A00612
GENSA00612EA 705 EUR
GENSA00612
Anti-His tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Tag
Antigen: His tag (His tag)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR