Anti-GST Mouse Monoclonal Antibody

Lieferant: Genscript
A00865-100 A00865-200 A00865-500
GENSA00865-100EA 302 EUR
GENSA00865-100 GENSA00865-200 GENSA00865-500
Anti-GST Mouse Monoclonal Antibody
Antikörper
Type: Tag
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR