Mouse IgG Isotype Control (Unconjugated)

Lieferant: Genscript
A01007
GENSA01007EA 138 EUR
GENSA01007
Mouse IgG Isotype Control (Unconjugated)
Antikörper
Type: Control
Antigen: IgG (Mouse) (mouse IgG)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR