Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Genscript
A01296
GENSA01296EA 516 EUR
GENSA01296
Anti-HA tag Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR