Anti-GFP Rabbit polyclonal antibody unconjugated

Lieferant: Genscript
A01388-40
GENSA01388-40EA 173 EUR
GENSA01388-40
Anti-GFP Rabbit polyclonal antibody unconjugated
Antikörper
Type: Tag
Antigen: GFP (green fluorescent protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR