Anti-His tag Mouse Monoclonal Antibody (FITC (Fluorescein))

Lieferant: Genscript
A01620
GENSA01620EA 705 EUR
GENSA01620
Anti-His tag Mouse Monoclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Tag
Antigen: His tag (His tag)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR