Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (FITC (Fluorescein))

Lieferant: Genscript
A01632
GENSA01632EA 260 EUR
GENSA01632
Anti-DYKDDDDK Tag Mouse Monoclonal Antibody (FITC (Fluorescein))
Antikörper
Type: Tag
Antigen: DYKDDDDK Tag (DYKDDDDK tag)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR