Anti-HSA Mouse Monoclonal Antibody [clone: 2C7B4]

Lieferant: Genscript
A01731-40
GENSA01731-40EA 192 EUR
GENSA01731-40
Anti-HSA Mouse Monoclonal Antibody [clone: 2C7B4]
Antikörper
Type: Primary
Antigen: HSA
Clonality: monoclonal
Clone: 2C7B4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR