Anti-TFRC Mouse Monoclonal Antibody

Lieferant: Merck Millipore (Oncogene)
GR08L-100
GR08L-100EA 569 EUR
GR08L-100
Anti-TFRC Mouse Monoclonal Antibody
Antikörper
Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR