Yeast transformation kit, Fast Yeast Transformation™
Lieferant: G Bioscienes
GZ-1
GZ-1EA
557
EUR
GZ-1
Yeast transformation kit, Fast Yeast Transformation™
Nukleinsäure-Reagenzien
Transformations-Reagenzien
Fast Yeast Transformation™ is a rapid single step yeast transformation kit that takes less than 10 minutes to prepare competent yeast cells. The competent yeast cells can be used immediately or frozen for later use. This method is suitable for both circular and linear plasmid transformations.
- High transformation efficiency: 10⁵ to 10⁶ transformants/μg circular DNA
- Broad spectrum: Compatible with C. albicans, S. pombe, P. pastoris, or S. cerevisiae
- Transformation procedure takes less than an hour
- Frozen competent cells are good for use for up to 6 months
- Simple protocol for multiple plasmid transformation
- Generates plasmids suitable for multiple downstream applications