Lauryl tryptose broth MPN tubes
Lieferant: Hach
2217515
HACH2217515EA
68
EUR
HACH2217515
Lauryl tryptose broth MPN tubes
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
Lauryl tryptose broth tubes with MUG microbiological media, single strength.
- Save time and labour costs
- Ready to use MPN tubes
- With minimal training requirements