Lauryl tryptose broth MPN tubes
Lieferant: Hach
2217515
HACH2217515EA 68 EUR
HACH2217515
Lauryl tryptose broth MPN tubes
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
Lauryl tryptose broth tubes with MUG microbiological media, single strength.
  • Save time and labour costs
  • Ready to use MPN tubes
  • With minimal training requirements