Media set
Lieferant: Hach
2277700
HACH2277700EA
60.4
EUR
HACH2277700
Media set
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
Media set is a colorimetric method that is used for determination of total bacteria.
- ToxTrak™ Toxicity test speeds sample evaluations
- Economically alternative to bioassays
It includes dehydrated tryptic soy broth, screw cap vial and sterile transfer pippets.