Media set
Lieferant: Hach
2277700
HACH2277700EA 60.4 EUR
HACH2277700
Media set
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
Media set is a colorimetric method that is used for determination of total bacteria.
  • ToxTrak™ Toxicity test speeds sample evaluations
  • Economically alternative to bioassays
It includes dehydrated tryptic soy broth, screw cap vial and sterile transfer pippets.