Settle plates, single-bagged
Lieferant: MERCK LIFE SCIENCE
1460040020
1460020020
1460020120
1460030120
HEIP1460040020EA
80
EUR
HEIP1460040020
HEIP1460020020
HEIP1460020120
HEIP1460030120
Settle plates, single-bagged
Medien für die Mikrobiologie
Fertigmedien für die Mikrobiologie
A wide range of ready-to-use culture media for the environmental monitoring. 90 mm LI (Long Incubation) plates for the determination of the total cell count and for the test for specified microorganisms
- Designed for the environmental monitoring in non critical areas