Settle plates, single-bagged
Lieferant: MERCK LIFE SCIENCE
1460040020 1460020020 1460020120 1460030120
HEIP1460040020EA 80 EUR
HEIP1460040020 HEIP1460020020 HEIP1460020120 HEIP1460030120
Settle plates, single-bagged
Medien für die Mikrobiologie Fertigmedien für die Mikrobiologie
A wide range of ready-to-use culture media for the environmental monitoring. 90 mm LI (Long Incubation) plates for the determination of the total cell count and for the test for specified microorganisms
  • Designed for the environmental monitoring in non critical areas