Capillary tubing kits
Lieferant: Agilent
5067-4601
HEWL5067-4601EA 929 EUR
HEWL5067-4601
Capillary tubing kits
Schläuche Chromatographie-Schläuche
Kit includes all capillaries and fittings, as well as bulk PEEK capillaries and a capillary cutter to add maximum flexibility.
  • Solvent selection kit- flow rate upto 10 ml/min