Capillaries for InfinityLab quick turn fittings
Lieferant: Agilent
5500-1188 5500-1189 5500-1190 5500-1191 5500-1192 5500-1193 5500-1194 5500-1195 5500-1196 5500-1197 5500-1198 5500-1200 5500-1205 5500-1206 5500-1232 5500-1233 5500-1234 5500-1235 5500-1236 5500-1237 5500-1261 5500-1262 5500-1263
558-0661EA 56 EUR
558-0661 558-0662 558-0663 558-0664 558-0665 558-0666 558-0667 558-0668 558-0669 558-0670 558-0671 HEWL5500-1200 HEWL5500-1205 HEWL5500-1206 558-0674 558-0675 558-0676 558-0677 558-0678 558-0679 558-0683 558-0684 558-0685
Capillaries for InfinityLab quick turn fittings
Schläuche Chromatographie-Schläuche
These capillaries are available in different lengths and internal diameters, including 0,075 mm ID, 0,12 mm ID, 0,17 mm ID and 0,25 mm ID to fit most UHPLC as well as HPLC applications.
  • Made of stainless steel
The 0,25 mm ID capillaries with a female connection are optimised for Waters Alliance systems.